Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FARP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106231 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-231 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-231 Supplier Abnova Corporation Supplier No. H00010160B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FARP1 protein.

This gene was originally isolated through subtractive hybridization due to its increased expression in differentiated chondrocytes versus dedifferentiated chondrocytes. The resulting protein contains a predicted ezrin-like domain, a Dbl homology domain, and a pleckstrin homology domain. It is believed to be a member of the band 4.1 superfamily whose members link the cytoskeleton to the cell membrane. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: MGEIEQRPTPGSRLGAPENSGISTLERGQKPPPTPSGKLVSIKIQMLDDTQEAFEVPMVSSSSFLKAIGSSWTGWVLRCSMKPKHHSHLIEKFGEDRILTHLTGSISYTNWAGSRSLAVTVTEELLNLF

Specifications

Antigen FARP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FARP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene FARP1
Gene Accession No. NM_001001715.1
Gene Alias CDEP/MGC87400/PLEKHC2
Gene Symbols FARP1
Host Species Mouse
Immunogen FARP1 (NP_001001715.1,1 a.a. ∽ 129 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10160
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.