Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAS, Mouse, Clone: 2R2, Abnova™

Catalog No. 89111382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-382 Supplier Abnova Corporation Supplier No. MAB3378
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against FAS.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contains a death domain. It has been shown to play a central role in the physiological regulation of programmed cell death, and has been implicated in the pathogenesis of various malignancies and diseases of the immune system. The interaction of this receptor with its ligand allows the formation of a death-inducing signaling complex that includes Fas-associated death domain protein (FADD), caspase 8, and caspase 10. The autoproteolytic processing of the caspases in the complex triggers a downstream caspase cascade, and leads to apoptosis. This receptor has been also shown to activate NF-kappaB, MAPK3/ERK1, and MAPK8/JNK, and is found to be involved in transducing the proliferating signals in normal diploid fibroblast and T cells. At least eight alternatively spliced transcript variants have been described, some of which are candidates for nonsense-mediated decay (NMD). The isoforms lacking the transmembrane domain may negatively regulate the apoptosis mediated by the full length isoform. [provided by RefSeq

Specifications

Antigen FAS
Applications Flow Cytometry
Classification Monoclonal
Clone 2R2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against FAS.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene FAS
Gene Alias ALPS1A/APO-1/APT1/CD95/FAS1/FASTM/TNFRSF6
Gene Symbols FAS
Host Species Mouse
Immunogen Mouse L cells stably over-expressing human Fas.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 355
Test Specificity This antibody is specific to human Fas.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.