Learn More
FAT1, Mouse, Clone: Fat-1, Abnova™
Mouse monoclonal antibody raised against FAT1.
Supplier: Abnova Corporation MAB7207
Description
This gene is an ortholog of the Drosophila fat gene, which encodes a tumor suppressor essential for controlling cell proliferation during Drosophila development. The gene product is a member of the cadherin superfamily, a group of integral membrane proteins characterized by the presence of cadherin-type repeats. In addition to containing 34 tandem cadherin-type repeats, the gene product has five epidermal growth factor (EGF)-like repeats and one laminin A-G domain. This gene is expressed at high levels in a number of fetal epithelia. Its product probably functions as an adhesion molecule and/or signaling receptor, and is likely to be important in developmental processes and cell communication. Transcript variants derived from alternative splicing and/or alternative promoter usage exist, but they have not been fully described. [provided by RefSeq
Specifications
FAT1 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
FAT1 | |
FAT1 | |
Cytoplasmic domain of drosphila FAT1. | |
RUO | |
2195 | |
Drosophila | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
Fat-1 | |
Mouse monoclonal antibody raised against FAT1. | |
In PBS | |
CDHF7/FAT/ME5/hFat1 | |
Mouse | |
100 μg | |
Primary | |
FAT (Fat-1). Expected to cross-react with Rat (85% identity with immunogen) and Chimpanzee (92% identity with immunogen) due to sequence homology. Predicted molecular weight 506 KDa. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.