Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBLN5, Mouse, Clone: 3F8D5, Abnova™

Catalog No. 89136019 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-019 Supplier Abnova Corporation Supplier No. MAB5582
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant FBLN5.

Specifications

Antigen FBLN5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F8D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant FBLN5.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to FBLN5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.