Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL22, Mouse, Clone: 1B6, Abnova™

Catalog No. 89107749 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-749 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-749 Supplier Abnova Corporation Supplier No. H00283807M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FBXL22.

Members of the F-box protein family, such as FBXL22, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MHITQLNRECLLHLFSFLDKDSRKSLARTCSQLHDVFEDPALWSLLHFRSLTELQKDNFLLGPALRSLSICWHSSRVQVCSIEDWLKSAFQRSICSRHESLVNDFLLRVC

Specifications

Antigen FBXL22
Applications ELISA
Classification Monoclonal
Clone 1B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FBXL22.
Formulation ascites with no preservative
Gene FBXL22
Gene Accession No. NM_203373
Gene Alias FLJ39626/Fbl22/MGC75496
Gene Symbols FBXL22
Host Species Mouse
Immunogen FBXL22 (NP_976307,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283807
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.