Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO11, Mouse, Clone: 4C12, Abnova™

Catalog No. 89107367 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-367 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-367 Supplier Abnova Corporation Supplier No. H00080204M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FBXO11.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: KAVSRGQCLYKISSYTSYPMHDFYRCHTCNTTDRNAICVNCIKKCHQGHDVEFIRHDRFFCDCGAGTLSNPCTLAGEPTHDTDTLYDSAPPIESNTLQHN

Specifications

Antigen FBXO11
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FBXO11.
Formulation ascites with no preservative
Gene FBXO11
Gene Accession No. NM_025133
Gene Alias FBX11/FLJ12673/MGC44383/PRMT9/UBR6/UG063H01/VIT1
Gene Symbols FBXO11
Host Species Mouse
Immunogen FBXO11 (NP_079409,744 a.a. ∽ 843 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80204
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.