Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO16, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107661 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-661 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-661 Supplier Abnova Corporation Supplier No. H00157574A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FBXO16.

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbx class. [provided by RefSeq]

Sequence: MMAFAPPKNTDGPKMQTKMSTWTPLNHQLLNDRVFEERRALLGKWFDKWTDSQRRRILTGLLERCSLSQQKFCCRKLQEKIPAEALDFTTKLPRVLSLYI

Specifications

Antigen FBXO16
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXO16.
Formulation 50% glycerol
Gene FBXO16
Gene Accession No. NM_172366
Gene Alias FBX16/MGC125923/MGC125924/MGC125925
Gene Symbols FBXO16
Host Species Mouse
Immunogen FBXO16 (NP_758954, 1 a.a. to 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157574
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.