Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO34, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-002 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-002 Supplier Abnova Corporation Supplier No. H00055030A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FBXO34.

Members of the F-box protein family, such as FBXO34, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MHLKPYWKLQKKEHPPEVSRETQRTPMNHQKAVNDETCKASHITSSVFPSASLGKASSRKPFGILSPNVLCSMSGKSPVESSLNVKTKKNAPSATIHQGE

Specifications

Antigen FBXO34
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FBXO34.
Formulation 50% glycerol
Gene FBXO34
Gene Accession No. NM_017943
Gene Alias CGI-301/DKFZp547C162/FLJ20725/Fbx34/MGC126434/MGC126435
Gene Symbols FBXO34
Host Species Mouse
Immunogen FBXO34 (NP_060413, 1 a.a. to 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55030
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.