Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCER2, Mouse, Clone: Tu1, Abnova™

Catalog No. 89113488 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-488 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-488 Supplier Abnova Corporation Supplier No. MAB8550
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length native FCER2.

The human leukocyte differentiation antigen CD23 (FCE2) is a key molecule for B-cell activation and growth. It is the low-affinity receptor for IgE. The truncated molecule can be secreted, then functioning as a potent mitogenic growth factor.[supplied by OMIM

Specifications

Antigen FCER2
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone Tu1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native FCER2.
Dilution Flow cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene FCER2
Gene Alias CD23/CD23A/CLEC4J/FCE2/IGEBF
Gene Symbols FCER2
Host Species Mouse
Immunogen Native purified FCER2 from human Tonsil lymphocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2208
Test Specificity This antibody is specific to FCEP2.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.