Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR1A, Mouse, Clone: 10.1, Abnova™

Catalog No. 89112178 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-178 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-178 Supplier Abnova Corporation Supplier No. MAB5230

Mouse monoclonal antibody raised against native FCGR1A.

This gene encodes a protein that plays an important role in the immune response. This protein is a high-affinity Fc-gamma receptor. The gene is one of three related gene family members located on chromosome 1. [provided by RefSeq

Specifications

Antigen FCGR1A
Applications Flow Cytometry
Classification Monoclonal
Clone 10.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native FCGR1A.
Dilution Flow Cytometry (10 ul to label 106 cells in 100 ul) The optimal working dilution should be determined by the end user.
Formulation In 200mM Tris-HCl pH 7.4 (0.09% sodium azide)
Gene FCGR1A
Gene Alias CD64/CD64A/FCRI/FLJ18345/IGFR1
Gene Symbols FCGR1A
Host Species Mouse
Immunogen Native purified FCGR1A from human monocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Test Specificity This antibody reacts with the human CD64 cell surface antigen, a 75 kD glycoprotein expressed by monocytes. The antigen acts as a high affinity receptor for human IgG, and is also known as FcRI. This antibody blocks binding of immunoglobulin to FcRI.
Target Species Baboon, Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.