Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR1C, Mouse, PE, Clone: 10.1, Abnova™

Catalog No. 89108451 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-451 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-451 Supplier Abnova Corporation Supplier No. MAB10246
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of FCGR1A.

This gene encodes a protein that plays an important role in the immune response. This protein is a high-affinity Fc-gamma receptor. The gene is one of three related gene family members located on chromosome 1. [provided by RefSeq

Specifications

Antigen FCGR1C
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10.1
Conjugate PE
Description Mouse monoclonal antibody raised against of FCGR1A.
Dilution Flow Cytometry (10 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene FCGR1A
Gene Alias CD64/CD64A/FCRI/FLJ18345/IGFR1
Gene Symbols FCGR1A
Host Species Mouse
Immunogen Rheumatoid synovial fluid cells and fibronectin purified human monocytes.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2209
Test Specificity FCGR1A, a single chain type I glycoprotein, that is expressed on monocytes/macrophages, dendritic cells, and activated granulocytes.
Target Species Human, Non-human Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.