Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-FCGR2A/C, Clone: 11B6,

Catalog No. MABF841 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF841 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF841 Supplier MilliporeSigma™ Supplier No. MABF841
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FCGR2A/C clone: 11B6 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting

FcγRI/CD64, FcγRII/CD32, FcγRIII/CD16, and FcγRIV/CD16-2 represent the four known classes of IgG Fc receptors (FcγRs). FcγRII, FcγRIII, and FcγRIV are low-affinity receptors for monomeric IgG, whereas FcγRI is the only high-affinity FcγR. FcγRs play important roles in inflammatory cell activation, clearance, presentation of Ag, and maintenance of IgG homeostasis. In addition to binding immune complex (IC), FcγRs have been shown to bind non-IgG ligands. For example, FcγRII is shown to bind oxLDL, while FcγRIII binding to an E. coli component is reported to negatively regulate the function of macrophage receptor with collagenous structure (MARCO). FcyRII activation is also reported to transduce a negative regulatory signal against CD38 cross-linking-induced proliferation of resting mature B cells. Low affinity immunoglobulin gamma Fc region receptor II-a/b/c (IgG Fc receptor II-a/-b/-c, CDw32, Fc-gamma RII-a/-b/-c, Fc-gamma-RIIa/b/c, FcRII-a/-b/-c, CD32) are encoded by the FCGR2A/B/C (CD32, FCG2, IGFR2) genes in human (UniProt P12318, P31994, P31995; Gene ID 2212, 2213, 9103). The FcγRIIs encoded by the three genes are highly conserved in their extracellular portion, with significant variation in the transmembrane and intracellular regions. All three types of transcripts alternative splicing to generate additional isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCGR2A/C
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 11B6
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P12318; P31995
Gene Symbols CD32; FCG2; FCGR2A; FCGR2A1; FCGR2B; FCGR2C; IGFR2
Host Species Mouse
Immunogen Linear peptide corresponding to a cytoplasmic domain sequence identical in human FcγRIIa and FcγRIIc.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001129691.1; NP_963857.3
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.