Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR3B, Mouse, Clone: MEM-154, Abnova™

Catalog No. 89108210 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-210 Supplier Abnova Corporation Supplier No. MAB0879
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native FCGR3B.

Specifications

Antigen FCGR3B
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-154
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native FCGR3B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 8.0 (0.09% sodium azide)
Gene FCGR3B
Gene Alias CD16/CD16b/FCG3/FCGR3
Gene Symbols FCGR3B
Host Species Mouse
Immunogen Native purified FCGR3B from human granulocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2215
Test Specificity This antibody reacts with the epitope on CD16 antigen that residing in proximity to FG loop (probably BC or C'E loop). This antibody reacts with CD16+ granulocytes.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.