Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCN3, Mouse, Clone: 4B4, Abnova™

Catalog No. 89105884 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-884 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-884 Supplier Abnova Corporation Supplier No. H00008547M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FCN3.

Ficolins are a group of proteins which consist of a collagen-like domain and a fibrinogen-like domain. In human serum, there are two types of ficolins, both of which have lectin activity. The protein encoded by this gene is a thermolabile beta-2-macroglycoprotein found in all human serum and is a member of the ficolin/opsonin p35 lectin family. The protein, which was initially identified based on its reactivity with sera from patients with systemic lupus erythematosus, has been shown to have a calcium-independent lectin activity. The protein can activate the complement pathway in association with MASPs and sMAP, thereby aiding in host defense through the activation of the lectin pathway. Alternative splicing occurs at this locus and two variants, each encoding a distinct isoform, have been identified. [provided by RefSeq

Sequence: MDLLWILPSLWLLLLGGPACLKTQEHPSCPGPRELEASKVVLLPSCPGAPGSPGEKGAPGPQGPPGPPGKMGPKGEPGPRNCRELLSQGATLSGWYHLCLPEGRALPVFCDMDTEGGGWLVFQRRQDGSVDFFRSWSSYRAGFGNQESEFWLGNENLHQLTLQGNWELRVELEDFNGNRTFAHYATFRLLGEVDHYQLALGKFSEGTAGDSLSLHSGRPLTTYDADHDSSNSNCAVIVHGAWWYASCYRSNLNGRYAVSEAAAHKYGIDWASGRGVGHPYRRVRMMLR

Specifications

Antigen FCN3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4B4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant FCN3.
Formulation ascites with no preservative
Gene FCN3
Gene Accession No. BC020731
Gene Alias FCNH/HAKA1/MGC22543
Gene Symbols FCN3
Host Species Mouse
Immunogen FCN3 (AAH20731,1 a.a. ∽ 288 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8547
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.