Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin, Mouse, Clone: 4G8C3, Abnova™

Catalog No. 89110500 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-500 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-500 Supplier Abnova Corporation Supplier No. MAB1568
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length native ferritin.

Specifications

Antigen Ferritin
Applications ELISA
Classification Monoclonal
Clone 4G8C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native ferritin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene Alias MGC71996
Gene Symbols FTL
Host Species Mouse
Immunogen Native purified ferritin from human liver.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2512/2495
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.