Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-FGF-23, Clone: FGF23/638, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | FGF-23 |
---|---|
Clone | FGF23/638 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1-5ug/ml for coating, Functional |
Applications | ELISA, Functional Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23461702
![]() |
Novus Biologicals
NBP2346170.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for ELISA, Functional.Specifications
FGF-23 | |
1.0 mg/mL | |
ELISA, Functional Assay | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer | |
10mM PBS - BSA free with No Preservative | |
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
FGF23 | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
FGF23/638 | |
ELISA 1-5ug/ml for coating, Functional | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9GZV9 | |
8074 | |
Recombinant human FGF23 protein | |
Primary | |
Protein A purified | |
Fibroblast growth factor-1 (FGF-1), also designated acidic FGF, and fibroblast growth factor-2 (FGF-2), also designated basic FGF, are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin. Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10 through FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure, and are capable of transforming cultured cells when overexpressed in trans- fected cells. Cellular receptors for FGFs are members of a second multigene family, including four tyrosine kinases designated Flg (FGFR-1), Bek (FGFR-L), TKF and FGFR-3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title