Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF3, Mouse, Clone: MSD1, Abnova™

Catalog No. 89113124 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-124 Supplier Abnova Corporation Supplier No. MAB7094
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of FGF3.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was identified by its similarity with mouse fgf3/int-2, a proto-oncogene activated in virally induced mammary tumors in the mouse. Frequent amplification of this gene has been found in human tumors, which may be important for neoplastic transformation and tumor progression. Studies of the similar genes in mouse and chicken suggested the role in inner ear formation. [provided by RefSeq

Sequence: RRTQKSSLFLPRVL

Specifications

Antigen FGF3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MSD1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of FGF3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene FGF3
Gene Alias HBGF-3/INT2
Gene Symbols FGF3
Host Species Mouse
Immunogen A synthetic peptide (conjugated with BSA) corresponding to human FGF3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2248
Test Specificity Detects a band at 27 KDa.
Target Species Human, Mouse, Xenopus species
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.