Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibrillin, Mouse, Clone: 11C1.3, Abnova™
Mouse monoclonal antibody raised against native Fibrillin.
Supplier: Abnova Corporation MAB3218
Specifications
Fibrillin | |
Monoclonal | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Bovine, Human | |
Liquid |
Immunofluorescence, Immunohistochemistry, Western Blot | |
11C1.3 | |
Mouse monoclonal antibody raised against native Fibrillin. | |
In 100mM BBS, pH 8.2 | |
Native purified bovine Fibrillin. | |
RUO | |
human and bovine fibrillin (Mr 350 KDa). | |
Store at 4°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction