Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen, Mouse, Clone: 27C8, Abnova™

Catalog No. 89111168 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-168 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-168 Supplier Abnova Corporation Supplier No. MAB2946
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Fibrinogen.

Specifications

Antigen Fibrinogen
Applications ELISA, Western Blot
Classification Monoclonal
Clone 27C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Fibrinogen.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias Fib2/MGC119422/MGC119423/MGC119425
Gene Symbols FGA
Host Species Mouse
Immunogen Human Fibrinogen.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2243/2244/2266
Test Specificity This antibody reacts with human fibrinogen and fibrin degradation products.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.