Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Surface Protein, Mouse, Clone: 1B10, Abnova™

Catalog No. 89136356 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-356 Supplier Abnova Corporation Supplier No. MAB7784

Mouse monoclonal antibody raised against native Fibroblast Surface Protein.

Specifications

Antigen Fibroblast Surface Protein
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 1B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Fibroblast Surface Protein.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Fibroblast Surface Protein from human thymus.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes an epitope on primary cultured human fibroblasts, fibroblast cell lines, tissue macrophages and 95% of peripheral blood monocytes. It does not react with vascular smooth muscle cells. This antibody may be used for removal of fibroblasts from human cultured cells by inhibition of fibroblast adherence to the culture vessel or by cytotoxic effect in the presence of rabbit complement.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.