Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Fibronectin/Anastellin, Clone: 2755-8, Azide Free, Novus Biologicals™
SDP

Catalog No. p-5326300 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23452602 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23452602 Supplier Novus Biologicals Supplier No. NBP2345260.2 MG

Mouse Monoclonal Antibody

Fibronectin Monoclonal antibody specifically detects Fibronectin in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen Fibronectin
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2755-8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml, CyTOF-ready
Formulation 10mM PBS - BSA free with No Preservative
Gene Accession No. P02751
Gene Symbols FN1
Host Species Mouse
Immunogen T-cell lymphoma biopsy
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2335
Test Specificity Fibronectin is an extracellular matrix glycoprotein present on most cell surfaces, in extracellular fluids and in plasma. A high molecular weight heterodimeric protein, it was originally discovered as a protein missing from the surfaces of virus-transformed cells, and it has been shown to be involved in various functions including cell adhesion, cell motility and wound healing. Alternative splicing and glycosylation give rise to several different forms of Fibronectin, some of which exhibit restricted tissue distribution or association with malignancies. It has been shown that Myofibroblasts phenotype formation correlates with the occurrence of glycosylated Fibronectin and Fibronectin splice variants in Dupuytren's disease
Reconstitution Protein A purified
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.