Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIGF, Mouse, Clone: 2E79, Abnova™

Catalog No. 89111484 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-484 Supplier Abnova Corporation Supplier No. MAB3582

Mouse monoclonal antibody raised against recombinant FIGF.

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C. [provided by RefSeq

Specifications

Antigen FIGF
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Clone: 2E79
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant FIGF.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from purified antibody without addition stabilizers
Gene FIGF
Gene Alias VEGF-D/VEGFD
Gene Symbols FIGF
Host Species Mouse
Immunogen Recombinant protein corresponding to human FIGF.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2277
Test Specificity This antibody reacts with human VEGFD, no reactivity with VEGFA and VEGFB.
Target Species Human
Content And Storage Store at -20°C on dry atmosphere.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.