Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIS1, Mouse, Clone: 1G9, Abnova™

Catalog No. 89106847 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-847 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-847 Supplier Abnova Corporation Supplier No. H00051024M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FIS1.

The balance between fission and fusion regulates the morphology of mitochondria. TTC11 is a component of a mitochondrial complex that promotes mitochondrial fission (James et al., 2003 [PubMed 12783892]).[supplied by OMIM

Sequence: MEAVLNELVSVEDLLKFEKKFQSEKAAGSVSKSTQFEYAWCLVRSKYNDDIRKGIVLLEELLPKGSKEEQRDYVFYLAVGNYRLKEYEKALKYVRGLLQTEPQNNQAKELERLIDKAMKKDGLVGMAIVGGMALGVAGLAGLIGLAVSKSKS

Specifications

Antigen FIS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant FIS1.
Formulation ascites with no preservative
Gene FIS1
Gene Accession No. BC003540
Gene Alias CGI-135/TTC11
Gene Symbols FIS1
Host Species Mouse
Immunogen FIS1 (AAH03540.1,1 a.a. ∽ 152 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51024
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.