Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FITC isomer 1, Mouse, Clone: NI 239, Abnova™

Catalog No. 89134764 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-764 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-764 Supplier Abnova Corporation Supplier No. MAB10079
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against FITC.

Specifications

Antigen FITC isomer 1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone NI 239
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against FITC.
Dilution ELISA (1:500) Western Blot (1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.2
Host Species Mouse
Immunogen Purified FITC isomer 1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antiserum has not been tested for cross-reactivity with other fluorescent compounds.
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 0.5mL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.