Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP11, Mouse, Clone: 1213s, Abnova™

Catalog No. 89113594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-594 Supplier Abnova Corporation Supplier No. MAB8809
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant FKBP11.

FKBP11 belongs to the FKBP family of peptidyl-prolyl cis/trans isomerases, which catalyze the folding of proline-containing polypeptides. The peptidyl-prolyl isomerase activity of FKBP proteins is inhibited by the immunosuppressant compounds FK506 and rapamycin (Rulten et al., 2006 [PubMed 16596453]).[supplied by OMIM

Specifications

Antigen FKBP11
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1213s
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant FKBP11.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH 7.0 (0.02% Proclin 300)
Gene FKBP11
Gene Alias FKBP19/MGC54182
Gene Symbols FKBP11
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 28-201 of human FKBP11.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51303
Test Specificity This antibody recognizes recombinant protein FKBP11.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.