Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP4, Mouse, ATTO488, Clone: Hi52C, Abnova™

Catalog No. 89109535 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-535 Supplier Abnova Corporation Supplier No. MAB11374
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of FKBP4.

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It has high structural and functional similarity to FK506-binding protein 1A (FKBP1A), but unlike FKBP1A, this protein does not have immunosuppressant activity when complexed with FK506. It interacts with interferon regulatory factor-4 and plays an important role in immunoregulatory gene expression in B and T lymphocytes. This encoded protein is known to associate with phytanoyl-CoA alpha-hydroxylase. It can also associate with two heat shock proteins (hsp90 and hsp70) and thus may play a role in the intracellular trafficking of hetero-oligomeric forms of the steroid hormone receptors. This protein correlates strongly with adeno-associated virus type 2 vectors (AAV) resulting in a significant increase in AAV-mediated transgene expression in human cell lines. Thus this encoded protein is thought to have important implications for the optimal use of AAV vectors in human gene therapy. The human genome contains several nontranscribed pseudogenes similar to this gene. [provided by RefSeq

Specifications

Antigen FKBP4
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Hi52C
Conjugate Atto 488
Description Mouse monoclonal antibody raised against synthetic peptide of FKBP4.
Dilution Western Blot (1:2000) Immunohistochemistry (1:250) Immunoprecipitation (5 ug with 10-20 uL Protein A beads) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene FKBP4
Gene Alias FKBP52/FKBP59/HBI/Hsp56/PPIase/p52
Gene Symbols FKBP4
Host Species Mouse
Immunogen A synthetic peptide corresponding to residues of human FKBP4.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2288
Target Species Canine, Hamster, Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.