Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLG, Mouse, Clone: FLG01, Abnova™

Catalog No. 89111461 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-461 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-461 Supplier Abnova Corporation Supplier No. MAB3538

Mouse monoclonal antibody raised against recombinant FLG.

Profilaggrin is a major protein component of the keratohyalin granules of mammalian epidermis. It is initially expressed as a large polyprotein precursor which is subsequently proteolytically processed into individual functional filaggrin molecules. The filaggrins show wide species variations and their aberrant expression has been implicated in a number of keratinizing disorders (Baden et al., 1974 [PubMed 4132224]; Holbrook et al., 1982 [PubMed 6174530]; Sybert et al., 1985 [PubMed 2579164]).[supplied by OMIM

Specifications

Antigen FLG
Applications Immunohistochemistry
Classification Monoclonal
Clone FLG01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant FLG.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (BSA, 0.09% sodium azide)
Gene FLG
Gene Alias ATOD2
Gene Symbols FLG
Host Species Mouse
Immunogen Recombinant protein corresponding to human FLG.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2312
Test Specificity This antibody reacts with human filaggrin 38 KDa and pro-filaggrin (∼ 400 KDa).
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.