Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ35767, Mouse, Clone: 2F3, Abnova™

Catalog No. 89107796 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-796 Supplier Abnova Corporation Supplier No. H00400629M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FLJ35767.

Sequence: MCPPVSMRYEEEGMSYLYASWMYQLQHGDQLSICFTCFKAAFLDFKDLLESEDWEEDNWDPELMEHTEAESEQEGSSGMELSWGQSPGQPVQGGSEAWGPGTLAAAPEGLEDAGLDPHFVPTELWPQEAVPLGLGLEDADWTQGLPWRFEELLTCSHWPSFFPS

Specifications

Antigen FLJ35767
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant FLJ35767.
Formulation PBS with no preservative; pH 7.4
Gene FLJ35767
Gene Accession No. NM_207459.1
Gene Symbols FLJ35767
Host Species Mouse
Immunogen FLJ35767 (NP_997342.1,1 a.a. ∽ 164 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400629
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.