Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ39155, Mouse, Clone: 1H3, Abnova™

Catalog No. 89107614 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-614 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-614 Supplier Abnova Corporation Supplier No. H00133584M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FLJ39155.

Sequence: DGRWHRVKAVRDGQSGKITVDDYGARTGKSPGMMRQLNINGALYVGGMKEIALHTNRQYMRGLVGCISHFTLSTDYHISLVEDAVDGK

Specifications

Antigen FLJ39155
Applications ELISA
Classification Monoclonal
Clone 1H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FLJ39155.
Formulation ascites with no preservative
Gene EGFLAM
Gene Accession No. NM_152403
Gene Alias AGRINL/AGRNL/FLJ39155
Gene Symbols EGFLAM
Host Species Mouse
Immunogen FLJ39155 (NP_689616,914 a.a. ∽ 1001 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133584
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.