Learn More
FN1, Mouse, Clone: 568, Abnova™
Mouse monoclonal antibody raised against native FN1.
Supplier: Abnova Corporation MAB3537
Description
This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq
Specifications
| FN1 | |
| Monoclonal | |
| Unconjugated | |
| In phosphate buffer, pH 7.3 (0.8% BSA, 0.09% sodium azide) | |
| CIG, DKFZp686F10164, DKFZp686H0342, DKFZp686I1370, DKFZp686O13149, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF | |
| Mouse | |
| 1 mL | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry | |
| 568 | |
| fibronectin 1 | |
| FN1 | |
| FN1 | |
| Native purified FN1 from human cultivated fibroblasts. | |
| RUO | |
| 2335 | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.