Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FN1, Mouse, Clone: DH1, Abnova™

Catalog No. 89112203 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-203 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-203 Supplier Abnova Corporation Supplier No. MAB5281
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native FN1.

This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq

Specifications

Antigen FN1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone DH1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native FN1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene FN1
Gene Alias CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF
Gene Symbols FN1
Host Species Mouse
Immunogen Native purified human FN1 from A8387 fibrosarcoma cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2335
Test Specificity This antibody is specific to extradomain A (EDA) sequence of a cellular fibronectin and recognizes thus only the cellular fibronectin. Clone DHI has been shown to specifically block chondrocyte condensation in chicken embryos. The monoclonal antibody DH1 recognizes only cellular form of Fibronectin.
Target Species Chicken, Guinea Pig, Human, Rabbit, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.