Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLH1, Mouse, PE, Clone: GCP-05, Abnova™

Catalog No. 89112891 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-891 Supplier Abnova Corporation Supplier No. MAB6477
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against FOLH1.

This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants. [provided by RefSeq

Specifications

Antigen FOLH1
Applications Flow Cytometry
Classification Monoclonal
Clone GCP-05
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against FOLH1.
Dilution Flow Cytometry (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene FOLH1
Gene Alias FGCP/FOLH/GCP2/GCPII/NAALAD1/NAALAdase/PSM/PSMA/mGCP
Gene Symbols FOLH1
Host Species Mouse
Immunogen Amino acids 44-750 of human FOLH1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2346
Test Specificity This antibody ecognizes extracellular domain of glutamate carboxy peptidase II (NAALADase, FOLH1, PSMA), an approximately 95-110 KDa transmembrane glycoprotein expressed mainly in tumour neovasculatures, nervous system and jejunum, which is animportant prostate tumour marker.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.