Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Foot-and-mouth disease (FMDV), Mouse, Clone: 1H4, Abnova™
Mouse monoclonal antibody raised against Foot-and-mouth disease (FMDV).
Supplier: Abnova Corporation MAB8526
This item is not returnable.
View return policy
Specifications
Foot-and-mouth disease (FMDV) | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Virus | |
Liquid |
ELISA | |
1H4 | |
Mouse monoclonal antibody raised against Foot-and-mouth disease (FMDV). | |
In PBS, pH 7.4 (0.09% sodium azide, 0.1% gelatin) | |
FMD virus. | |
RUO | |
Specific for NS- non-structural proteins. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction