Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Fox1, Clone: 1D10,

Catalog No. MABE985MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE985MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE985MI Supplier MilliporeSigma™ Supplier No. MABE985
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Fox1 clone: 1D10 in Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

RNA binding protein fox-1 homolog 1 (UniProt Q9JJ43; also known as Ataxin-2-binding protein 1, Fox-1 homolog A, Hexaribonucleotide binding protein 1) is encoded by the Rbfox1 (also known as A2bp, A2bp1, Fox-1, Hrnbp1) gene (ORF MNCb-3035; Gene ID 268859) in murine species. The RNA-binding Fox (Rbfox) family of splicing factors is comprised of three members, Rbfox1 (Fox-1 or A2BP1), Rbfox2 (Fox-2 or RBM9), and Rbfox3 (Fox-3, HRNBP3 or NeuN). Rbfox proteins regulate splicing of many neuronal transcripts (pre-mRNAs) by binding the sequence (U)GCAUG in introns flanking alternative exons. A (U)GCAUG motif that lies downstream of the alternative exon generally promotes Rbfox-dependent exon inclusion, whereas an upstream motif will usually repress exon inclusion. Rbfox1 transcript itself is subjected to alternative splicing in response to chronic cell depolarization. This results in the increased expression of the nuclear, splicing-active Rbfox1 isoform, which in turn leads to increased splicing of Rbfox1 target transcripts of proteins, including N-methyl D-aspartate (NMDA) receptor 1 (Grin1) and a calcium ATPase (Atp2b1), that play important roles in neuronal excitation and calcium homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fox1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1D10
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9JJ43
Gene Symbols Rbfox1; A2bp; A2bp1; Fox1; MNCb-3035
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to mouse Fox1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_067452
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.