Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXA3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104657 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-657 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-657 Supplier Abnova Corporation Supplier No. H00003171B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FOXA3 protein.

This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved. [provided by RefSeq

Sequence: MLGSVKMEAHDLAEWSYYPEAGEVYSPVTPVPTMAPLNSYMTLNPLSSPYPPGGLPASPLPSGPLAPPAPAAPLGPTFPGLGVSGGSSSSGYGAPGPGLVHGKEMPKGYRRPLAHAKPPYSYISLITMAIQQAPGKMLTLSEIYQWIMDLFPYYRENQQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWALHPSSGNMFENGCYLRRQKRFKLEEKVKKGGSGAATTTRNGTGSAASTTTPAATVTSPPQPPPPAPEPEAQGGEDVGALDCGSPASSTPYFTGLELPGELKLDAPYNFNHPFSINNLMSEQTPAPPKLDVGFGGYGAEGGEPGVYYQGLYSRSLLNAS

Specifications

Antigen FOXA3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FOXA3 protein.
Formulation No additive
Gene FOXA3
Gene Accession No. NM_004497.2
Gene Alias FKHH3/HNF3G/MGC10179/TCF3G
Gene Symbols FOXA3
Host Species Mouse
Immunogen FOXA3 (NP_004488.2, 1 a.a. to 350 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3171
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.