Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXD3 Mouse anti-Human, Monkey, Mouse, Clone: 5G9, Abnova™

Catalog No. 89108850 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-850 Supplier Abnova Corporation Supplier No. MAB10652
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant FOXD3.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1. [provided by RefSeq

Specifications

Antigen FOXD3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant FOXD3.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene FOXD3
Gene Alias AIS1/Genesis/HFH2
Gene Symbols FOXD3
Host Species Mouse
Immunogen Recombinant protein corresponding to human FOXD3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27022
Target Species Human, Monkey, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.