Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxp3, Mouse, Clone: 3G3, Abnova™

Catalog No. 89107875 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-875 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-875 Supplier Abnova Corporation Supplier No. MAB0107
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant Foxp3.

Specifications

Antigen Foxp3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3G3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Foxp3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene Foxp3
Gene Alias JM2/scurfin/sf
Gene Symbols Foxp3
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length mouse Foxp3.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20371
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.