Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxp3, Mouse, PE, Clone: 3G3, Abnova™

Catalog No. 89113814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-814 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-814 Supplier Abnova Corporation Supplier No. MAB9852
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant Foxp3.

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone 3G3
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against full length recombinant Foxp3.
Dilution Flow Cytometry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Gene Foxp3
Gene Alias JM2/scurfin/sf
Gene Symbols Foxp3
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length mouse Foxp3.
Purification Method Size-exclusion chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20371
Test Specificity The mouse monoclonal antibody 3G3 recognizes N-terminal region of Foxp3, a 47-55 kDa transcription factor, which is the master regulator in the development and function of regulatory T cells.
Target Species Human, Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.