Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXQ1 (M04J), Mouse anti-Human, Clone: 1A10, Abnova™

Catalog No. 89107519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-519 Supplier Abnova Corporation Supplier No. H00094234M04J
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FOXQ1.

FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis (Bieller et al., 2001 [PubMed 11747606]).[supplied by OMIM

Sequence: RSKPYTRRPKPPYSYIALIAMAIRDSAGGRLTLAEINEYLMGKFPFFRGSYTGWRNSVRHNLSLNDCFVKVLRDPSRPWGKDNYWMLNPNSEYTFADGVFRRRRKRLSHR

Specifications

Antigen FOXQ1
Applications ELISA
Classification Monoclonal
Clone 1A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FOXQ1.
Formulation PBS with no preservative; pH 7.4
Gene FOXQ1
Gene Accession No. NM_033260
Gene Alias HFH1
Gene Symbols FOXQ1
Host Species Mouse
Immunogen FOXQ1 (NP_150285,110 a.a. ∽ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94234
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.