Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXQ1 (M10C), Mouse anti-Human, Clone: 2G6, Abnova™

Catalog No. 89107522 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-522 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-522 Supplier Abnova Corporation Supplier No. H00094234M10C
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FOXQ1.

FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis (Bieller et al., 2001 [PubMed 11747606]).[supplied by OMIM

Sequence: RSKPYTRRPKPPYSYIALIAMAIRDSAGGRLTLAEINEYLMGKFPFFRGSYTGWRNSVRHNLSLNDCFVKVLRDPSRPWGKDNYWMLNPNSEYTFADGVFRRRRKRLSHR

Specifications

Antigen FOXQ1
Applications ELISA
Classification Monoclonal
Clone 2G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FOXQ1.
Formulation tissue culture supernatant with no preservative
Gene FOXQ1
Gene Accession No. NM_033260
Gene Alias HFH1
Gene Symbols FOXQ1
Host Species Mouse
Immunogen FOXQ1 (NP_150285,110 a.a. ∽ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94234
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.