Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-FPR1/2, Clone: NFPR1,

Catalog No. MABF271MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF271MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF271MI Supplier MilliporeSigma™ Supplier No. MABF271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FPR1/2 clone: NFPR1 in Human samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blotting

N-formy peptide receptor 1 (also known as N-formylpeptide chemoattractant receptor or fMet-Leu-Phe receptor) & 2 (also known as RFP, FMLP-R-I, Lipoxin A4 receptor, LXA4 receptor, FMLP-related receptor I, Formyl peptide receptor-like 1) are multi-pass cell membrane G protein-coupled receptors (GPCRs) encoded by the FPR1 (also known as FPR and FMLP) and FPR2 (also known as FPR2 and FMLP-R-II) gene in human, respectively. The FPR1 gene encodes the high-affinity, and FPR2 the low-affinity, receptor for N-formyl-methionyl peptides (FMLP). Both receptors mediate FMLP-induced activation of neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPR1/2
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NFPR1
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P21462
Gene Symbols FPR1
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to the FPR1 (305-GQDFRERLI-313) and FPR2 (306- GQDFRERLI-314) of human FPR1/2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001180235
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.