Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-FPR1, C-Term Dephospho-, Clone: NFPRb,

Catalog No. MABF287MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF287MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF287MI Supplier MilliporeSigma™ Supplier No. MABF287
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FPR1, C-Term Dephospho- clone: NFPRb in Human samples, and it is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting

fMet-Leu-Phe receptor (UniProt P21462; also known as fMLP receptor, FPR, N-formylpeptide chemoattractant receptor, N-formyl peptide receptor) is encoded by the FPR1 (also known as FMLP) gene in human (Entrez Gene ID 2357). N-formyl-methionyl peptides (FMLPs) are powerful neutrophils chemotactic factors. FMLPs exert their biological activies via binding G-protein-coupled N-formyl peptide chemoattractant receptors (FPRs). Occupancy of FPR1 by FMLPs is shown to induce regulatory phosphorylation of cytoplasmic serine/threonine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPR1, C-Term Dephospho-
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone NFPRb
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P21462
Gene Symbols FPR1
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to human FPR1 near the C-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001180235
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.