Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRAT1, Mouse, Clone: 7C9, Abnova™

Catalog No. 89106198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-198 Supplier Abnova Corporation Supplier No. H00010023M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FRAT1.

The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq

Sequence: RLQQRRGSQPETRTGDDDPHRLLQQLVLSGNLIKEAVRRLHSRRLQLRAKLPQRPLLGPLSAPVHEPPSPRSPRAACSDPGASGRAQLRTGDGVLVPGS

Specifications

Antigen FRAT1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FRAT1.
Formulation PBS with no preservative; pH 7.4
Gene FRAT1
Gene Accession No. NM_005479.3
Gene Alias FLJ97193
Gene Symbols FRAT1
Host Species Mouse
Immunogen FRAT1 (NP_005470.2,181 a.a. ∽ 279 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10023
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.