Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRG1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104515 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-515 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-515 Supplier Abnova Corporation Supplier No. H00002483B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FRG1 protein.

This gene maps to a location 100kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy (FSHD). It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus. [provided by RefSeq

Sequence: MAEYSYVKSTKLVLKGTKTKSKKKKSKDKKRKREEDEETQLDIVGIWWTVTNFGEISGTIAIEMDKGTYIHALDNGLFTLGAPHKEVDEGPSPPEQFTAVKLSDSRIALKSGYGKYLGINSDGLVVGRSDAIGPREQWEPVFQNGKMALLASNSCFIRCNEAGDIEAKSKTAGEEEMIKIRSCAERETKKKDDIPEEDKGNVKQCEINYVKKFQSFQDHKLKISKEDSKILKKARKDGFLHETLLDRRAKLKADRYCK

Specifications

Antigen FRG1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FRG1 protein.
Formulation No additive
Gene FRG1
Gene Accession No. NM_004477
Gene Alias FRG1A/FSG1
Gene Symbols FRG1
Host Species Mouse
Immunogen FRG1 (NP_004468, 1 a.a. to 258 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2483
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.