Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSBA, Mouse, Clone: 2F5/5, Abnova™

Catalog No. 89136347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-347 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-347 Supplier Abnova Corporation Supplier No. MAB7754

Mouse monoclonal antibody raised against FSBA.

Specifications

Antigen FSBA
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2F5/5
Conjugate Unconjugated
Formulation In PBS
Host Species Mouse
Immunogen Fluorylsulponylbenzoyla.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity FSBA 2F5/5 has been shown to detect Protein Kinase C.
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.