Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSH, Mouse, Clone: 2H42F9, Abnova™

Catalog No. 89110506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-506 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-506 Supplier Abnova Corporation Supplier No. MAB1577
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length native FSH.

The pituitary glycoprotein hormone family includes follicle-stimulating hormone, luteinizing hormone, chorionic gonadotropin, and thyroid-stimulating hormone. All of these glycoproteins consist of an identical alpha subunit and a hormone-specific beta subunit. This gene encodes the beta subunit of follicle-stimulating hormone. In conjunction with luteinizing hormone, follicle-stimulating hormone induces egg and sperm production. Alternative splicing results in two transcript variants encoding the same protein. [provided by RefSeq

Specifications

Antigen FSH
Applications ELISA
Classification Monoclonal
Clone 2H42F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length native FSH.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Gene FSHB
Gene Symbols FSHB
Host Species Mouse
Immunogen Native purified human FSH.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2488
Test Specificity This antibody react with FSH, and shows low cross-reactivity (less than 0.02%) against hCG, LH, TSH.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.