Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSTL3, Mouse, Clone: 1G12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FSTL3.
Supplier: Abnova Corporation H00010272M12
Description
Follistatin-like 3 is a secreted glycoprotein of the follistatin-module-protein family. It may have a role in leukemogenesis. [provided by RefSeq
Sequence: MGSGNPAPGGVCWLQQGQEATCSLVLQTDVTRAECCASGNIDTAWSNLTHPGNKINLLGFLGLVHCLPCKDSCDGVECGPGKACRMLGGRPRCECAPDCSGLPARLQVCGSDGATYRDECELRAARCRGHPDLSVMYRGRCRKSCEHVVCPRPQSCVVDQTGSAHCVVCRAAPCPVPSSPGQELCGNNNVTYISSCHMRQATCFLGRSIGVRHAGSCAGTPEEPPGGESAEEEENFVSpecifications
FSTL3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005860.2 | |
FSTL3 | |
FSTL3 (NP_005851.1,27 a.a. ∽ 263 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
1G12 | |
Mouse monoclonal antibody raised against a full-length recombinant FSTL3. | |
FSTL3 | |
FLRG/FSRP | |
Mouse | |
Protein A Purification | |
RUO | |
10272 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction