Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fto, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89136426 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-426 Supplier Abnova Corporation Supplier No. MAB8175
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Fto.

Specifications

Antigen Fto
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Fto.
Dilution Western Blot (1:1000) Immunofluorescence (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA)
Host Species Mouse
Immunogen A synthetic peptide corresponding to Fto.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody has been directly tested for reactivity in human, mouse and rat. The immunogen sequence has 100% homology with bovine, canine, horse, non-human primate and sheep. specific to the ∼58 KDa Fto protein in Western blots of rat testes lysate. This antibody has also been used for immunocytochemistry with neuronal progenitor cells.
Target Species Bovine, Canine, Equine, Human, Mouse, Non-human Primate, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.