Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUBP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105967 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-967 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-967 Supplier Abnova Corporation Supplier No. H00008880A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant FUBP1.

This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the noncoding strand. This protein has been shown to function as an ATP-dependent DNA helicase. [provided by RefSeq

Sequence: VNDAFKDALQRARQIAAKIGGDAGTSLNSNDYGYGGQKRPLEDGDQPDAKKVAPQNDSFGTQLPPMHQQQSRSVMTEEYKVPDGMVGFIIGRGGEQISRIQQESGCKIQI

Specifications

Antigen FUBP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant FUBP1.
Formulation 50% glycerol
Gene FUBP1
Gene Accession No. NM_003902
Gene Alias FBP/FUBP
Gene Symbols FUBP1
Host Species Mouse
Immunogen FUBP1 (NP_003893, 27 a.a. to 136 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8880
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.