Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUT4, Mouse, Biotin, Clone: TG1, Abnova™

Catalog No. 89112269 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-269 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-269 Supplier Abnova Corporation Supplier No. MAB5499
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against FUT4.

The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq

Specifications

Antigen FUT4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone TG1
Conjugate Biotin
Description Mouse monoclonal antibody raised against FUT4.
Dilution Flow Cytometry (using 1ug to stain 1 x 106 cells) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Gene FUT4
Gene Alias CD15/ELFT/FCT3A/FUC-TIV/FUTIV
Gene Symbols FUT4
Host Species Mouse
Immunogen Thymocyte glycoprotein fractions adsorbed to Con A and eluted with 0.1M glucose.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2526
Test Specificity Recognizes a (Mr 156 KDa) cell surface glycoprotein expressed on virtually all peripheral blood granulocytes. The CD15 antigen is expressed on approximately 95% of human peripheral blood eosinophils and neutrophils and is present on approximately 20% of mature circulating monocytes. Anti-CD15 has virtually no reactivity with T or B cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.